Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206924.1 | NP_001193853.1 | 6 | 1215 | ||
NM_001206925.1 | NP_001193854.1 | 6 | 1501 | |||
NM_006889.4 | NP_008820.3 | 7 | 1606 | |||
NM_175862.4 | NP_787058.4 | 7 | 1551 | |||
NM_176892.1 | NP_795711.1 | 6 | 1462 | |||
GenBank mRNA | AK294663.1 | - | 6 | 1210 | ||
AK316203.1 | - | 6 | 1496 | |||
BC040261.1 | - | 7 | 1538 | 5173789 |