Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004182.3 | NP_004173.1 | 7 | 571 | ||
NM_153477.2 | NP_705582.1 | 6 | 749 | |||
NR_045559.1 | 5 | 681 | ||||
NR_045560.1 | 8 | 636 | ||||
GenBank mRNA | AF083241.1 | - | 7 | 537 | ||
AF083242.1 | - | 6 | 687 | |||
AF092737.1 | - | 7 | 552 | |||
AK223320.1 | - | 7 | 525 | |||
BC000720.1 | - | 6 | 506 | 3505415 | ||
BC008890.1 | - | 6 | 506 | 3959862 | ||
BM471483.1 | - | 5 | 609 | |||
BQ681820.1 | - | 3 | 245 |