Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000947.4 | NP_000938.2 | 9 | 901 | ||
XM_011514731.2 | XP_011513033.1 | 3 | 2668 | |||
XM_017011004.1 | XP_016866493.1 | 10 | 1093 | |||
XM_017011005.1 | XP_016866494.1 | 10 | 1093 | |||
XM_017011006.1 | XP_016866495.1 | 10 | 1093 | |||
XM_017011007.1 | XP_016866496.1 | 7 | 748 | |||
GenBank mRNA | AK291915.1 | - | 9 | 888 | ||
BC064931.1 | - | 9 | 886 | 6148494 | ||
X74331.1 | - | 9 | 894 |