Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016184.3 | NP_057268.1 | 4 | 651 | ||
NM_194447.2 | NP_919429.2 | 3 | 534 | |||
NM_194448.2 | NP_919430.1 | 2 | 435 | |||
NM_194450.2 | NP_919432.1 | 3 | 552 | |||
XM_011520684.2 | XP_011518986.1 | 3 | 1912 | |||
XM_017019382.1 | XP_016874871.1 | 5 | 572 | |||
GenBank mRNA | AF067800.1 | - | 4 | 672 | ||
AF109146.1 | - | 4 | 498 | |||
AF200738.1 | - | 3 | 305 | |||
AF328684.1 | - | 4 | 643 | |||
AJ133532.1 | - | 4 | 645 | |||
BC074822.2 | - | 4 | 426 | 30915322 | ||
BC074874.2 | - | 4 | 426 | 30915470 | ||
BC117439.1 | - | 4 | 546 | 40125990 | ||
BC117441.1 | - | 4 | 546 | 40125992 |