Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000851.3 | NP_000842.2 | 6, 7 | 527 | ||
XM_005270784.4 | XP_005270841.1 | 7, 8 | 553 | |||
XM_005270785.4 | XP_005270842.1 | 3, 4 | 268 | |||
GenBank mRNA | AK125820.1 | - | 1, 2 | 1499 | ||
AK127250.1 | - | 4, 5 | 1239 | |||
AK289673.1 | - | 6, 7 | 484 | |||
AK315260.1 | - | 6, 7 | 478 | |||
BC058881.1 | - | 6, 7 | 504 | 6193461 | ||
BI824480.1 | - | 6, 7 | 527 | |||
DB089682.1 | - | 6, 7 | 527 | |||
L02321.1 | - | 6, 7 | 513 |