Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016190.2 | NP_057274.1 | 3 | 281 | ||
GenBank mRNA | AF077831.1 | - | 3 | 259 | ||
AK291301.1 | - | 3 | 270 | |||
AK298640.1 | - | 3 | 270 | |||
AK316568.1 | - | 3 | 270 | |||
BC030807.1 | - | 3 | 270 | 4271624 | ||
DA420219.1 | - | 3 | 281 |