Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS18C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297769.1 | NP_001284698.1 | 2, 3 | 298 | ||
NM_016067.3 | NP_057151.1 | 2, 3 | 298 | |||
GenBank mRNA | AF151892.1 | - | 2, 3 | 211 | ||
AK026620.1 | - | 2, 3 | 265 | |||
BC005186.1 | - | 2, 3 | 189 | 3839381 | ||
BE874586.1 | - | 2, 3 | 140 |