Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270523.1 | NP_001257452.1 | 3 | 304 | ||
NM_001270524.1 | NP_001257453.1 | 2 | 457 | |||
NM_001270525.1 | NP_001257454.1 | 1 | 227 | |||
NM_021728.3 | NP_068374.1 | 3 | 304 | |||
NM_172337.2 | NP_758840.1 | 1 | 227 | |||
NR_073034.1 | 3 | 304 | ||||
NR_073036.1 | 1 | 227 | ||||
GenBank mRNA | AB593056.1 | - | 1 | 230 | ||
AB593058.1 | - | 3 | 304 | |||
AF093138.1 | - | 1 | 20 | |||
AK314271.1 | - | 2 | 360 | |||
BC032579.1 | - | 3 | 305 | 5493541 | ||
BE781530.1 | - | 1 | 191 | |||
BU176852.1 | - | 1 | 210 | |||
CV812961.1 | - | 3 | 281 | |||
DB294481.1 | - | 3 | 307 |