Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NETO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201465.1 | NP_001188394.1 | 2, 3 | 359 | ||
NM_138966.3 | NP_620416.1 | 2, 3 | 359 | |||
NM_138999.1 | NP_620552.1 | 2, 3 | 174 | |||
XM_005266774.4 | XP_005266831.1 | 2, 3 | 297 | |||
XM_017026017.1 | XP_016881506.1 | 2, 3 | 221 | |||
XM_017026018.1 | XP_016881507.1 | 2, 3 | 221 | |||
XM_017026019.1 | XP_016881508.1 | 2, 3 | 381 | |||
XM_017026020.1 | XP_016881509.1 | 2, 3 | 385 | |||
XM_017026021.1 | XP_016881510.1 | 1, 2 | 47 | |||
XM_017026022.1 | XP_016881511.1 | 2, 3 | 139 | |||
XM_017026023.1 | XP_016881512.1 | 2, 3 | 139 | |||
XM_017026024.1 | XP_016881513.1 | 2, 3 | 354 | |||
GenBank mRNA | AF448838.1 | - | 2, 3 | 365 | ||
AF448839.1 | - | 2, 3 | 174 | |||
BC050329.1 | - | 2, 3 | 361 | 4821317 |