Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005752.1 | NP_001005752.1 | 2 | 762 | ||
NM_024009.2 | NP_076872.1 | 2 | 1219 | |||
GenBank mRNA | AF052692.1 | - | 1 | 1737 | ||
AK026139.1 | - | 2 | 1218 | |||
AK312890.1 | - | 2 | 918 | |||
BC012918.1 | - | 2 | 751 | 4500186 | ||
BC110640.1 | - | 1 | 1794 | 4780218 |