Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010974.2 | NP_001010974.1 | 3 | 410 | ||
NM_017843.4 | NP_060313.3 | 3 | 410 | |||
NM_198799.3 | NP_942094.2 | 3 | 410 | |||
XM_011528886.2 | XP_011527188.1 | 3 | 333 | |||
XM_011528887.2 | XP_011527189.1 | 3 | 333 | |||
XM_017027932.1 | XP_016883421.1 | 3 | 333 | |||
GenBank mRNA | AF361220.1 | - | 3 | 410 | ||
AJ511266.1 | - | 3 | 234 | |||
AK000502.1 | - | 3 | 273 | |||
BC038381.1 | - | 3 | 260 | 4472674 | ||
BC047337.1 | - | 3 | 239 | |||
BP212127.1 | - | 3 | 374 |