Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRELP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002725.3 | NP_002716.1 | 3 | 3582 | ||
NM_201348.1 | NP_958505.1 | 3 | 3572 | |||
GenBank mRNA | BG569381.1 | - | 1 | 279 | ||
BM705293.1 | - | 1 | 318 |