Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAV2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206747.1 | NP_001193676.1 | 3 | 547 | ||
NM_001206748.1 | NP_001193677.1 | 2 | 432 | |||
NM_001233.4 | NP_001224.1 | 3 | 547 | |||
NM_198212.2 | NP_937855.1 | 2 | 359 | |||
GenBank mRNA | AA883287.1 | - | 2 | 177 | ||
AB451285.1 | - | 3 | 366 | |||
AB451417.1 | - | 3 | 366 | |||
AF035752.1 | - | 3 | 386 | |||
AK310786.1 | - | 2 | 220 | |||
AK311964.1 | - | 3 | 389 | |||
AY353255.1 | - | 3 | 378 | |||
BC005256.1 | - | 3 | 547 | 3829012 | ||
BT007051.1 | - | 3 | 366 | |||
CR407685.1 | - | 3 | 366 | |||
DA882274.1 | - | 2 | 432 | |||
DQ656068.1 | - | 14 | 1828 |
Gene Symbol: CAV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF179102.1 | - | 7 | 693 |
Gene Symbol: LOC105375463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | EF070114.1 | - | 11 | 1068 | ||
EF070118.1 | - | 5 | 468 |