Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319294.1 | NP_001306223.1 | 6 | 638 | ||
NM_001319295.1 | NP_001306224.1 | 6 | 710 | |||
NM_016121.4 | NP_057205.2 | 6 | 638 | |||
XM_005273158.1 | XP_005273215.1 | 6 | 904 | |||
GenBank mRNA | AK296275.1 | - | 4 | 171 | ||
AK316272.1 | - | 2 | 39 | |||
BC013868.2 | - | 6 | 625 | |||
BC094879.1 | - | 6 | 587 | 3078971 | ||
BC117188.1 | - | 6 | 520 | 40125739 | ||
BC126173.1 | - | 6 | 520 | 8991889 |