Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D3P1-DHX40P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002924.3 | 13 | 1529 |
Gene Symbol: RNFT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016125.3 | NP_057209.3 | 2 | 472 | ||
GenBank mRNA | AF100745.1 | - | 2 | 480 | ||
AK294806.1 | - | 2 | 453 | |||
AK295162.1 | - | 2 | 428 | |||
AK296197.1 | - | 2 | 453 | |||
BC006971.1 | - | 2 | 450 | |||
BC088365.1 | - | 2 | 435 | 30390062 | ||
DB456985.1 | - | 2 | 472 |