Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC729218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103825.1 | 3 | 927 | |||
NR_109983.1 | 10 | 1870 | ||||
GenBank mRNA | AK024248.1 | - | 10 | 1841 |
Gene Symbol: LINC00999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024497.2 | 8 | 1647 | |||
GenBank mRNA | AK128746.1 | - | 1 | 204 |
Gene Symbol: LINC01000 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024368.1 | 5 | 1174 | |||
GenBank mRNA | AK302487.1 | - | 5 | 1195 | ||
BC062355.1 | - | 5 | 1093 | |||
BC131589.1 | - | 5 | 1093 |
Gene Symbol: LINC01001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028326.1 | 3 | 963 | |||
GenBank mRNA | AK293878.1 | - | 3 | 991 |
Gene Symbol: LINC01061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037596.1 | 3 | 937 | |||
GenBank mRNA | AK294946.1 | - | 1 | 57 |
Gene Symbol: LINC01347 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029401.1 | 18 | 3613 | |||
GenBank mRNA | AL137733.1 | - | 18 | 3613 |
Gene Symbol: LOC100132062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028325.1 | 3 | 963 | |||
GenBank mRNA | BC112924.1 | - | 1 | 351 |
Gene Symbol: LOC100132287 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028322.1 | 3 | 963 | |||
GenBank mRNA | AK311126.1 | - | 1 | 137 |
Gene Symbol: LOC100133331 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028327.1 | 3 | 963 |
Gene Symbol: LOC101060341 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271793.4 | XP_005271850.1 | 3 | 892 | ||
XM_017012925.1 | XP_016868414.1 | 4 | 1096 | |||
XM_017012926.1 | XP_016868415.1 | 2 | 901 | |||
XM_017012927.1 | XP_016868416.1 | 3 | 913 |
Gene Symbol: LOC101929819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK302511.1 | - | 7 | 1443 |
Gene Symbol: LOC107986800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012928.1 | XP_016868417.1 | 3 | 858 | ||
XM_017012929.1 | XP_016868418.1 | 2 | 846 |
Gene Symbol: LOC729737 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039983.2 | 3 | 1286 | |||
GenBank mRNA | AK294489.1 | - | 1 | 619 | ||
AK302573.1 | - | 1 | 249 |