Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136540.1 | NP_001130012.1 | 2, 3 | 256 | ||
NM_003661.3 | NP_003652.2 | 2, 3 | 217 | |||
NM_145343.2 | NP_663318.1 | 3, 4 | 371 | |||
GenBank mRNA | AF019225.2 | - | 2, 3 | 216 | ||
AF305224.1 | - | 2, 3 | 156 | |||
AF305428.1 | - | 2, 3 | 196 | |||
AF323540.1 | - | 3, 4 | 370 | |||
AK296099.1 | - | 2, 3 | 166 | |||
AK309143.1 | - | 2, 3 | 214 | |||
AK313752.1 | - | 3, 4 | 310 | |||
BC017331.1 | - | 2, 3 | 147 | |||
BC112943.1 | - | 2, 3 | 172 | 4512933 | ||
BC113867.1 | - | 2, 3 | 147 | |||
BC114475.1 | - | 2, 3 | 147 | |||
BC127186.1 | - | 2, 3 | 147 | 40034503 | ||
BC141823.1 | - | 2, 3 | 172 | 40148156 | ||
BC142720.1 | - | 2, 3 | 172 | |||
BC143038.1 | - | 2, 3 | 147 | 40034505 | ||
BC143039.1 | - | 2, 3 | 147 | 40034506 | ||
BG419709.1 | - | 1, 2 | 86 | |||
DA375861.1 | - | 3, 4 | 312 | |||
DC340792.1 | - | 2, 3 | 194 | |||
KU178424.1 | - | 1, 2 | 49 |