Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014349.2 | NP_055164.1 | 4 | 1148 | ||
NM_030644.1 | NP_085147.1 | 5 | 1032 | |||
NM_145639.1 | NP_663614.1 | 4 | 919 | |||
NM_145640.2 | NP_663615.1 | 3 | 822 | |||
NM_145641.2 | NP_663616.1 | 5 | 2331 | |||
NM_145642.2 | NP_663617.1 | 4 | 2268 | |||
XM_006724324.1 | XP_006724387.1 | 5 | 910 | |||
XM_006724325.2 | XP_006724388.1 | 4 | 821 | |||
XM_017028946.1 | XP_016884435.1 | 4 | 748 | |||
XM_017028947.1 | XP_016884436.1 | 5 | 861 | |||
XM_017028948.1 | XP_016884437.1 | 6 | 1165 | |||
XM_017028949.1 | XP_016884438.1 | 6 | 1050 | |||
XM_017028950.1 | XP_016884439.1 | 5 | 1755 | |||
XM_017028951.1 | XP_016884440.1 | 4 | 1642 | |||
XM_017028952.1 | XP_016884441.1 | 3 | 668 | |||
GenBank mRNA | AF070675.1 | - | 4 | 988 | ||
AF305227.1 | - | 4 | 775 | |||
AK025349.1 | - | 4 | 745 | |||
AY014902.1 | - | 5 | 1032 | |||
AY014903.1 | - | 4 | 919 | |||
AY014904.1 | - | 3 | 822 | |||
AY014905.1 | - | 4 | 1148 | |||
AY014906.1 | - | 5 | 2331 | |||
AY014907.1 | - | 4 | 2268 | |||
BC042918.1 | - | 4 | 724 | 5932779 | ||
BC050596.1 | - | 4 | 779 | 6012757 | ||
CR456379.1 | - | 3 | 642 |