Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032546.3 | NP_115935.3 | 5, 6 | 1037 | ||
NM_187841.2 | NP_912730.2 | 4, 5 | 911 | |||
GenBank mRNA | AJ291714.1 | - | 5, 6 | 734 | ||
AK300699.1 | - | 5, 6 | 883 | |||
BC141807.1 | - | 4, 5 | 683 | 40147443 | ||
BF829678.1 | - | 1, 2 | 384 |