Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278791.1 | NP_001265720.1 | 8 | 925 | ||
NM_001278792.1 | NP_001265721.1 | 7 | 716 | |||
NM_001278793.1 | NP_001265722.1 | 7 | 823 | |||
NM_002914.4 | NP_002905.2 | 7 | 721 | |||
NM_181471.2 | NP_852136.1 | 8 | 823 | |||
XM_006716080.2 | XP_006716143.1 | 6 | 545 | |||
XM_017012492.1 | XP_016867981.1 | 8 | 801 | |||
XM_017012493.1 | XP_016867982.1 | 6 | 569 | |||
GenBank mRNA | AB451243.1 | - | 8 | 748 | ||
AB451368.1 | - | 8 | 748 | |||
BC002813.1 | - | 7 | 651 | 3637774 | ||
BM810551.1 | - | 7 | 628 | |||
BQ070526.1 | - | 8 | 773 | |||
BQ674131.1 | - | 7 | 675 | |||
BT007058.1 | - | 7 | 646 | |||
M87338.1 | - | 955 |