Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267721.1 | NP_001254650.1 | 4 | 669 | ||
NM_144603.3 | NP_653204.1 | 4 | 654 | |||
NM_172167.2 | NP_751907.1 | 4 | 657 | |||
NM_172168.2 | NP_751908.1 | 4 | 672 | |||
XM_005255099.4 | XP_005255156.1 | 5 | 1767 | |||
XM_017022927.1 | XP_016878416.1 | 5 | 1273 | |||
XM_017022928.1 | XP_016878417.1 | 5 | 630 | |||
GenBank mRNA | AB097667.1 | - | 4 | 467 | ||
AF532983.1 | - | 4 | 288 | |||
AF532984.1 | - | 4 | 291 | |||
AF532985.1 | - | 4 | 306 | |||
AF539796.1 | - | 4 | 287 | |||
AK292197.1 | - | 4 | 657 | |||
AK310198.1 | - | 2 | 859 | |||
AK313917.1 | - | 4 | 577 | |||
AY191359.1 | - | 4 | 303 | |||
AY255768.1 | - | 4 | 284 | |||
BC015917.2 | - | 4 | 544 | 4661469 |