Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000627.3 | NP_000618.3 | 30 | 5029 | ||
NM_001166264.1 | NP_001159736.1 | 29 | 4903 | |||
NM_001166265.1 | NP_001159737.1 | 30 | 4870 | |||
NM_001166266.1 | NP_001159738.1 | 29 | 4744 | |||
NM_206943.2 | NP_996826.2 | 34 | 5889 | |||
XM_005264317.3 | XP_005264374.1 | 34 | 5732 | |||
XM_005264318.3 | XP_005264375.1 | 33 | 5606 | |||
XM_011532853.2 | XP_011531155.1 | 34 | 6065 | |||
XM_011532855.2 | XP_011531157.1 | 34 | 5888 | |||
XM_011532856.2 | XP_011531158.1 | 33 | 5774 | |||
XM_011532857.2 | XP_011531159.1 | 33 | 5768 | |||
XM_011532858.2 | XP_011531160.1 | 33 | 5768 | |||
XM_011532859.2 | XP_011531161.1 | 34 | 5735 | |||
XM_011532860.2 | XP_011531162.1 | 33 | 5726 | |||
XM_011532861.2 | XP_011531163.1 | 32 | 5624 | |||
XM_017004108.1 | XP_016859597.1 | 33 | 5771 | |||
XM_017004109.1 | XP_016859598.1 | 33 | 5765 | |||
XM_017004110.1 | XP_016859599.1 | 32 | 5645 | |||
GenBank mRNA | AB208801.1 | - | 30 | 4828 | ||
M34057.1 | - | 30 | 4977 |