Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFATC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136021.2 | NP_001129493.1 | 3 | 1358 | ||
NM_001258292.1 | NP_001245221.1 | 3 | 1358 | |||
NM_001258294.1 | NP_001245223.1 | 3 | 844 | |||
NM_001258295.1 | NP_001245224.1 | 3 | 844 | |||
NM_001258296.1 | NP_001245225.1 | 3 | 922 | |||
NM_001258297.1 | NP_001245226.1 | 3 | 922 | |||
NM_012340.4 | NP_036472.2 | 3 | 1436 | |||
NM_173091.3 | NP_775114.1 | 3 | 1436 | |||
XM_011528824.2 | XP_011527126.1 | 3 | 1427 | |||
XM_011528825.2 | XP_011527127.1 | 3 | 1384 | |||
XM_011528826.2 | XP_011527128.1 | 3 | 865 | |||
XM_017027850.1 | XP_016883339.1 | 3 | 660 | |||
XM_017027851.1 | XP_016883340.1 | 3 | 1427 | |||
GenBank mRNA | AK226146.1 | - | 3 | 1358 | ||
AK304147.1 | - | 3 | 813 | |||
BC136418.1 | - | 3 | 1339 | 9020406 | ||
BC144074.1 | - | 3 | 1339 | 9052594 | ||
BM463165.1 | - | 227 | ||||
EU887573.1 | - | 3 | 1216 | |||
EU887574.1 | - | 3 | 1156 | |||
EU887575.1 | - | 3 | 844 | |||
EU887576.1 | - | 3 | 922 | |||
EU887577.1 | - | 3 | 1156 | |||
EU887578.1 | - | 3 | 1216 | |||
EU887579.1 | - | 3 | 844 | |||
EU887580.1 | - | 3 | 922 | |||
U43341.1 | - | 3 | 1436 | |||
U43342.1 | - | 3 | 1436 |