Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_110124.1 | 3 | 1083 | |||
NR_110132.1 | 3 | 573 | ||||
GenBank mRNA | D43770.1 | - | LIT1584 CMPD |
3 | 921 |