Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016060.2 | NP_057144.1 | 4 | 1015 | ||
GenBank mRNA | AF151883.1 | - | 4 | 987 |