Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243116.1 | NP_001230045.1 | 6 | 902 | ||
NM_002503.4 | NP_002494.2 | 6 | 1144 | |||
NR_040515.1 | 5 | 2159 | ||||
XM_006723226.2 | XP_006723289.2 | 6 | 1080 | |||
XM_006723227.3 | XP_006723290.3 | 6 | 1043 | |||
GenBank mRNA | AK057862.1 | - | 5 | 2110 | ||
BC015528.1 | - | 6 | 1075 | 3903794 | ||
BT006743.1 | - | 6 | 1041 | |||
BX111397.1 | - | 1 | 285 |