Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAGE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001187.1 | NP_001178.1 | 2 | 247 | ||
GenBank mRNA | AF527551.1 | - | 247 | |||
AF527552.1 | - | 247 | ||||
AF527553.1 | - | 2 | 247 | |||
AF527554.1 | - | 247 | ||||
BC107038.2 | - | 2 | 130 | 40002186 | ||
BC107039.2 | - | 2 | 150 | |||
BC113810.1 | - | 2 | 150 | |||
U19180.1 | - | 2 | 247 |
Gene Symbol: BAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182482.2 | NP_872288.2 | 2 | 255 | ||
GenBank mRNA | AF218570.1 | - | 2 | 255 | ||
AF527550.1 | - | 2 | 247 | |||
AK292540.1 | - | 2 | 295 | |||
BC101132.2 | - | 2 | 150 | |||
BC101133.2 | - | 2 | 150 |
Gene Symbol: BAGE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182481.1 | NP_872287.1 | 2 | 255 | ||
GenBank mRNA | AF339514.1 | - | 2 | 255 | ||
BC101134.2 | - | 2 | 150 | 40014666 |
Gene Symbol: BAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181704.1 | NP_859055.1 | 2 | 235 | ||
GenBank mRNA | AF339515.1 | - | 2 | 235 |
Gene Symbol: CCDC144NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004306.2 | NP_001004306.1 | 2, 3 | 543 | ||
XM_011523818.2 | XP_011522120.1 | 2, 3 | 990 | |||
XM_011523819.2 | XP_011522121.1 | 2, 3 | 961 | |||
XM_011523821.1 | XP_011522123.1 | 2, 3 | 162 | |||
GenBank mRNA | AW504417.1 | - | 2, 3 | 232 | ||
BC068597.1 | - | 2, 3 | 543 | 4837813 |