Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLOD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016080.3 | NP_057164.3 | 4 | 448 | ||
XM_006721540.2 | XP_006721603.1 | 4 | 470 | |||
XM_017024711.1 | XP_016880200.1 | 4 | 772 | |||
XM_017024712.1 | XP_016880201.1 | 4 | 687 | |||
GenBank mRNA | AF061730.1 | - | 4 | 393 | ||
AF151908.1 | - | 6 | 1191 | |||
AF177342.1 | - | 5 | 441 | |||
AF177343.1 | - | 4 | 424 | |||
AK001488.1 | - | 4 | 411 | |||
AK122678.1 | - | 2 | 936 | |||
AK226083.1 | - | 4 | 372 | |||
AK292315.1 | - | 4 | 373 | |||
AK296584.1 | - | 5 | 638 | |||
AK301751.1 | - | 4 | 325 | |||
BC008605.1 | - | 4 | 373 | 4186251 | ||
BC015848.1 | - | 4 | 401 | 4657199 |