Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBP1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033339.1 | 2 | 1639 | |||
GenBank mRNA | AK125775.1 | - | 3 | 1593 | ||
BC013775.1 | - | 1 | 742 |