Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR3DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322168.1 | NP_001309097.1 | 8 | 1206 | ||
NM_013289.2 | NP_037421.2 | 8 | 1206 | |||
GenBank mRNA | AF022049.1 | - | 8 | 1143 | ||
AF103010.1 | - | 8 | 1143 | |||
AF103011.1 | - | 8 | 1143 | |||
AF103012.1 | - | 8 | 1143 | |||
AF103013.1 | - | 8 | 1143 | |||
AF262968.1 | - | 6 | 1062 | |||
AF262969.1 | - | 6 | 1062 | |||
AF262970.1 | - | 6 | 1062 | |||
AF262971.1 | - | 6 | 1062 | |||
AF262972.1 | - | 6 | 1062 | |||
AF262973.1 | - | 8 | 1176 | |||
AF262974.1 | - | 8 | 1176 | |||
AJ417556.1 | - | 6 | 1092 | |||
AJ417557.1 | - | 6 | 1092 | |||
AY366236.1 | - | 8 | 1181 | |||
AY366249.1 | - | 8 | 1143 | |||
AY366250.1 | - | 8 | 1169 | |||
AY366251.1 | - | 8 | 1143 | |||
AY366252.1 | - | 8 | 1143 | |||
AY760050.1 | - | 8 | 1143 | |||
AY789065.1 | - | 8 | 1158 | |||
AY789066.1 | - | 8 | 1143 | |||
AY789072.1 | - | 8 | 1143 | |||
BC028206.1 | - | 8 | 1170 | 5215674 | ||
BC101540.1 | - | 8 | 1162 | 8069046 | ||
BC101542.1 | - | 8 | 1162 | 8069048 | ||
DQ178659.1 | - | 8 | 1143 | |||
EF462885.1 | - | 6 | 1073 | |||
EF472674.1 | - | 6 | 1073 | |||
EF472675.1 | - | 6 | 1073 | |||
EF472676.1 | - | 6 | 1073 | |||
EF472677.1 | - | 6 | 1073 | |||
EF472678.1 | - | 6 | 1073 | |||
EF472679.1 | - | 6 | 1073 | |||
L41269.1 | - | 8 | 1162 | |||
L76664.1 | - | 7 | 869 | |||
U30273.1 | - | 8 | 1145 | |||
U30274.1 | - | 8 | 1206 | |||
U31416.1 | - | 8 | 1168 | |||
U33328.1 | - | 7 | 1117 | |||
X94262.1 | - | 8 | 1144 |