Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040443.2 | NP_001035533.1 | 8 | 1082 | ||
NM_001040444.2 | NP_001035534.1 | 8 | 758 | |||
NR_135322.1 | 7 | 960 | ||||
NR_135323.1 | 9 | 894 | ||||
GenBank mRNA | AB011031.1 | - | 8 | 731 | ||
AF155105.1 | - | 8 | 702 | |||
AK026228.1 | - | 8 | 722 | |||
AK225986.1 | - | 9 | 844 | |||
BC017212.2 | - | 8 | 711 | 4054134 | ||
BX461254.2 | - | 7 | 590 |
Gene Symbol: SETDB2-PHF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320727.1 | NP_001307656.1 | 18 | 3078 | ||
NR_135324.1 | 18 | 3117 |