Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005743.1 | NP_001005743.1 | 4 | 359 | ||
NM_001005744.1 | NP_001005744.1 | 4 | 359 | |||
NM_001005745.1 | NP_001005745.1 | 4 | 359 | |||
NM_001320114.1 | NP_001307043.1 | 4 | 379 | |||
NM_003744.5 | NP_003735.3 | 4 | 359 | |||
GenBank mRNA | AF015040.1 | - | 4 | 317 | ||
AF108092.1 | - | 2 | 144 | |||
AF171938.1 | - | 4 | 309 | |||
AF171939.1 | - | 4 | 309 | |||
AF171940.1 | - | 4 | 309 | |||
AF171941.1 | - | 4 | 309 | |||
AK304193.1 | - | 2 | 162 | |||
AK315130.1 | - | 3 | 240 | |||
BC020788.1 | - | 3 | 240 | |||
BC068476.1 | - | 4 | 359 | 4822300 | ||
BX248073.1 | - | 3 | 216 | |||
BX248775.1 | - | 4 | 288 | |||
BX248781.1 | - | 3 | 254 | |||
BX647137.1 | - | 4 | 318 | |||
EU265734.1 | - | 2 | 78 | |||
EU265735.1 | - | 2 | 78 | |||
EU265736.1 | - | 2 | 78 | |||
EU265737.1 | - | 2 | 78 |