Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1LC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032514.3 | NP_115903.1 | 3 | 356 | ||
NM_181509.2 | NP_852610.1 | 4 | 302 | |||
XM_011529083.2 | XP_011527385.1 | 6 | 656 | |||
XM_011529084.2 | XP_011527386.1 | 6 | 657 | |||
XM_011529085.2 | XP_011527387.1 | 5 | 531 | |||
GenBank mRNA | AF276658.1 | - | 3 | 338 | ||
AL833855.1 | - | 3 | 249 | |||
BC015810.1 | - | 3 | 287 | 3909192 | ||
BM740344.1 | - | 4 | 331 | |||
BM919877.1 | - | 338 | ||||
BP212418.1 | - | 3 | 299 | |||
BT007452.1 | - | 3 | 184 | |||
EL954540.1 | - | 3 | 420 | |||
H18312.1 | - | 4 | 304 |