Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181481.4 | NP_852146.1 | 3 | 572 | ||
NM_181482.4 | NP_852147.1 | 4 | 831 | |||
XM_005258140.1 | XP_005258197.1 | 4 | 801 | |||
XM_006722353.1 | XP_006722416.1 | 5 | 858 | |||
XM_006722354.1 | XP_006722417.1 | 3 | 662 | |||
XM_017025965.1 | XP_016881454.1 | 7 | 1168 | |||
XM_017025966.1 | XP_016881455.1 | 6 | 1043 | |||
XM_017025967.1 | XP_016881456.1 | 6 | 1040 | |||
XM_017025968.1 | XP_016881457.1 | 5 | 947 | |||
GenBank mRNA | AF009424.1 | - | 3 | 575 | ||
AF009425.1 | - | 3 | 575 | |||
BC030199.1 | - | 4 | 765 | 5199354 |