Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001777.3 | NP_001768.1 | 11 | 1296 | ||
NM_198793.2 | NP_942088.1 | 9 | 1238 | |||
XM_005247908.1 | XP_005247965.1 | 10 | 1233 | |||
XM_005247909.1 | XP_005247966.1 | 8 | 1176 | |||
GenBank mRNA | AK124152.1 | - | 2 | 2147 | ||
AK289813.1 | - | 11 | 1237 | |||
AL832675.1 | - | 2 | 2953 | |||
BC010016.2 | - | 9 | 1140 | 4303534 | ||
BC012884.1 | - | 9 | 1161 | 3857911 | ||
BC037306.1 | - | 9 | 1175 | 5260986 | ||
X69398.1 | - | 11 | 1222 | |||
Z25521.1 | - | 9 | 1197 |