Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001777.3 | NP_001768.1 | 11 | 2371 | ||
NM_198793.2 | NP_942088.1 | 9 | 2313 | |||
XM_005247908.1 | XP_005247965.1 | 10 | 2308 | |||
XM_005247909.1 | XP_005247966.1 | 8 | 2251 | |||
GenBank mRNA | AL832675.1 | - | 2 | 4030 | ||
BC037306.1 | - | 9 | 2250 | 5260986 | ||
BC045593.1 | - | 1 | 940 | |||
BC053959.1 | - | 1 | 940 | |||
BX640889.1 | - | 1 | 940 |