Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002457.3 | NP_002448.3 | 15022 | |||
GenBank mRNA | L21998.1 | - | 15022 | |||
M94132.1 | - | 16 | 2410 |