Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000533.4 | NP_000524.3 | 7 | 2707 | ||
NM_001128834.2 | NP_001122306.1 | 8 | 2789 | |||
NM_001305004.1 | NP_001291933.1 | 7 | 2482 | |||
NM_199478.2 | NP_955772.1 | 7 | 2602 | |||
GenBank mRNA | AK128782.1 | - | 7 | 2630 | ||
AV731932.2 | - | 1 | 318 | |||
BC095452.1 | - | 7 | 2592 | 30717624 | ||
M27110.1 | - | 7 | 2625 |