Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015560.2 | NP_056375.2 | 2 | 404 | ||
NM_130831.2 | NP_570844.1 | 2 | 404 | |||
NM_130832.2 | NP_570845.1 | 2 | 404 | |||
NM_130833.2 | NP_570846.1 | 2 | 404 | |||
NM_130834.2 | NP_570847.2 | 2 | 404 | |||
NM_130835.2 | NP_570848.1 | 2 | 404 | |||
NM_130836.2 | NP_570849.2 | 2 | 404 | |||
NM_130837.2 | NP_570850.2 | 2 | 404 | |||
XM_017006500.1 | XP_016861989.1 | 3 | 379 | |||
GenBank mRNA | AB011139.1 | - | 2 | 225 | ||
BC075805.1 | - | 2 | 324 | 4214698 | ||
DA993450.1 | - | 1 | 73 |