Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171740.2 | NP_001165211.1 | 2 | 620 | ||
NM_001171741.2 | NP_001165212.1 | 2 | 415 | |||
NM_001171743.2 | NP_001165214.2 | 2 | 459 | |||
NM_016210.4 | NP_057294.2 | 3 | 709 | |||
XM_011533782.2 | XP_011532084.1 | 3 | 685 | |||
XM_011533783.2 | XP_011532085.1 | 2 | 575 | |||
XM_011533784.2 | XP_011532086.1 | 3 | 463 | |||
XM_011533785.2 | XP_011532087.1 | 2 | 401 | |||
XM_011533790.1 | XP_011532092.1 | 3 | 685 | |||
XM_017006545.1 | XP_016862034.1 | 3 | 685 | |||
XM_017006547.1 | XP_016862036.1 | 2 | 620 | |||
GenBank mRNA | AF188706.1 | - | 2 | 380 | ||
AK124438.1 | - | 2 | 460 | |||
AK308074.1 | - | 2 | 343 | |||
AK313128.1 | - | 3 | 583 | |||
AL833938.1 | - | 3 | 668 | |||
BC034766.1 | - | 2 | 493 | 5177784 |