Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013899.2 | NP_038927.2 | 3 | 490 | ||
GenBank mRNA | AF150090.1 | - | 3 | 467 | ||
AK008088.1 | - | 3 | 491 | |||
AK013546.1 | - | 3 | 422 | |||
BC031448.1 | - | 3 | 468 | 3257567 |