Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039141.2 | NP_001034230.1 | 15, 16 | 5963 | ||
NM_007032.5 | NP_008963.3 | 5, 6 | 557 | |||
NM_138632.2 | NP_619538.2 | 5, 6 | 557 | |||
GenBank mRNA | AF281030.1 | - | 5, 6 | 541 | ||
BC003618.2 | - | 5, 6 | 479 | |||
BC022200.1 | - | 5, 6 | 551 | |||
BQ880866.1 | - | 557 | ||||
BX342673.2 | - | 5, 6 | 551 | |||
DQ228003.1 | - | 13, 14 | 5626 | |||
DQ228005.1 | - | 14, 15 | 5828 |