Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128608.1 | NP_001122080.1 | 25 | 3086 | ||
NM_001265611.1 | NP_001252540.1 | 24 | 3068 | |||
NM_014994.2 | NP_055809.2 | 24 | 3068 | |||
NR_049761.1 | 23 | 2878 | ||||
NR_049762.1 | 22 | 2829 | ||||
XM_006720438.1 | XP_006720501.1 | 23 | 2885 | |||
XM_006720439.2 | XP_006720502.1 | 11 | 1160 | |||
XM_011521383.1 | XP_011519685.1 | 24 | 2903 | |||
XM_011521384.2 | XP_011519686.1 | 25 | 3050 | |||
XM_017022017.1 | XP_016877506.1 | 23 | 3179 | |||
GenBank mRNA | AB011168.2 | - | 25 | 2996 | ||
AB177850.1 | - | 16 | 1916 | |||
AK302482.1 | - | 22 | 2770 | |||
AL833267.1 | - | 24 | 2988 | |||
BC036660.1 | - | 23 | 2883 | 5266613 | ||
BC113983.1 | - | 24 | 2960 | 40083186 | ||
BC114493.1 | - | 25 | 2978 | 40083182 | ||
BX640787.1 | - | 20 | 3449 | |||
BX648786.1 | - | 24 | 3019 |