Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004113.5 | NP_004104.3 | 6 | 928 | ||
NM_021032.4 | NP_066360.1 | 5 | 1694 | |||
XM_005247227.2 | XP_005247284.1 | 5 | 892 | |||
XM_006713538.3 | XP_006713601.1 | 5 | 760 | |||
XM_017005879.1 | XP_016861368.1 | 5 | 779 | |||
GenBank mRNA | AK125307.1 | - | 5 | 1694 | ||
BC022524.1 | - | 6 | 912 | 4794600 |