Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLGA8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001023567.4 | NP_001018861.3 | 7 | 630 | ||
NR_027410.1 | 15 | 2878 | ||||
XM_011521596.1 | XP_011519898.1 | 14 | 1475 | |||
XM_011521597.1 | XP_011519899.1 | 13 | 1426 | |||
XM_017022210.1 | XP_016877699.1 | 15 | 1779 | |||
XM_017022211.1 | XP_016877700.1 | 15 | 1548 | |||
XM_017022212.1 | XP_016877701.1 | 14 | 2102 | |||
XM_017022213.1 | XP_016877702.1 | 14 | 1504 | |||
XM_017022214.1 | XP_016877703.1 | 15 | 1779 | |||
XM_017022215.1 | XP_016877704.1 | 15 | 2301 | |||
XM_017022216.1 | XP_016877705.1 | 14 | 2899 | |||
XM_017022217.1 | XP_016877706.1 | 13 | 2821 | |||
XM_017022218.1 | XP_016877707.1 | 15 | 2928 | |||
GenBank mRNA | AK097551.1 | - | 7 | 619 | ||
BC143254.1 | - | 7 | 576 | 9051756 | ||
BC144610.1 | - | 5 | 367 | 9053141 | ||
BX647673.1 | - | 14 | 2847 |
Gene Symbol: GOLGA8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181077.3 | NP_851422.1 | 7 | 630 | ||
NR_027409.1 | 15 | 2892 | ||||
XM_006720440.2 | XP_006720503.1 | 7 | 598 | |||
XM_017022018.1 | XP_016877507.1 | 11 | 3026 | |||
XM_017022019.1 | XP_016877508.1 | 10 | 2722 | |||
XM_017022020.1 | XP_016877509.1 | 5 | 720 | |||
GenBank mRNA | AB020662.1 | - | 6 | 610 | ||
AF163441.1 | - | 5 | 1054 | |||
AF204231.1 | - | 6 | 947 | |||
AL137686.1 | - | 5 | 354 | |||
BC032774.1 | - | 7 | 532 | |||
BC093673.1 | - | 7 | 576 | 7939518 | ||
BC104800.1 | - | 7 | 576 | 8143803 | ||
BC144609.1 | - | 5 | 367 | 9053140 | ||
BC150329.1 | - | 6 | 621 | |||
BC152410.1 | - | 6 | 610 | |||
BX648160.1 | - | 14 | 2846 | |||
BX648575.1 | - | 14 | 2599 | |||
CR749361.1 | - | 7 | 1783 |