Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 6 | 855 | ||
NM_001197317.2 | NP_001184246.1 | 4 | 662 | |||
NM_001197318.2 | NP_001184247.1 | 4 | 669 | |||
NM_001197319.2 | NP_001184248.1 | 3 | 558 | |||
NM_013269.5 | NP_037401.1 | 5 | 773 | |||
NR_036693.2 | 4 | 744 | ||||
GenBank mRNA | AF133299.1 | - | 5 | 759 | ||
AF285089.1 | - | 5 | 765 | |||
AK310150.1 | - | 7 | 1459 | |||
AK310554.1 | - | 4 | 735 | |||
BC019883.1 | - | 5 | 1770 | 4524891 |