Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289396.1 | NP_001276325.1 | 28 | 4248 | ||
NM_001289397.1 | NP_001276326.1 | 28 | 4087 | |||
NM_001289398.1 | NP_001276327.1 | 2 | 269 | |||
NM_001289399.1 | NP_001276328.1 | 3 | 256 | |||
NM_001289400.1 | NP_001276329.1 | 3 | 677 | |||
NM_003070.4 | NP_003061.3 | 28 | 4261 | |||
NM_139045.3 | NP_620614.2 | 28 | 4261 | |||
GenBank mRNA | AK094076.1 | - | 3 | 677 | ||
AK296373.1 | - | 28 | 3649 | |||
AK298045.1 | - | 3 | 256 | |||
AK299683.1 | - | 19 | 2454 | |||
AK300093.1 | - | 4 | 321 | |||
AK308941.1 | - | 2 | 414 | |||
AL558829.3 | - | 3 | 208 | |||
BI758293.1 | - | 2 | 270 | |||
CD107405.1 | - | 1 | 24 | |||
CD513892.1 | - | 1 | 65 | |||
CD702931.1 | - | 2 | 508 | |||
DA084762.1 | - | 2 | 266 | |||
DA823500.1 | - | 2 | 192 | |||
X72889.1 | - | 28 | 4249 |