Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098482.1 | NP_001091952.1 | 14, 15 | 1829 | ||
NM_015321.2 | NP_056136.2 | 13, 14 | 1781 | |||
XM_005259833.3 | XP_005259890.1 | 13, 14 | 1703 | |||
XM_005259834.1 | XP_005259891.1 | 12, 13 | 1688 | |||
XM_005259835.3 | XP_005259892.1 | 14, 15 | 1531 | |||
XM_005259836.3 | XP_005259893.1 | 13, 14 | 1426 | |||
XM_006722710.3 | XP_006722773.1 | 12, 13 | 1375 | |||
XM_011527842.2 | XP_011526144.1 | 13, 14 | 1685 | |||
GenBank mRNA | AB014516.1 | - | 14, 15 | 1694 | ||
AK024089.1 | - | 12, 13 | 1694 | |||
AY040323.1 | - | 12, 13 | 1694 | |||
AY360171.1 | - | 14, 15 | 1766 | |||
BC017075.2 | - | 9, 10 | 1262 | |||
BC023614.2 | - | 13, 14 | 1604 | |||
BC028050.1 | - | 13, 14 | 1713 | 4938995 |