Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037333.2 | NP_001032410.1 | 31 | 3983 | ||
NM_001291721.1 | NP_001278650.1 | 30 | 3905 | |||
NM_001291722.1 | NP_001278651.1 | 32 | 4058 | |||
NM_014376.3 | NP_055191.2 | 31 | 4109 | |||
XM_011534516.2 | XP_011532818.1 | 31 | 4249 | |||
XM_017009341.1 | XP_016864830.1 | 31 | 4098 | |||
XM_017009342.1 | XP_016864831.1 | 32 | 4020 | |||
GenBank mRNA | AB032994.2 | - | 32 | 3998 | ||
AF160973.1 | - | 31 | 3892 | |||
AK294220.1 | - | 26 | 3352 | |||
AK295865.1 | - | 28 | 3532 | |||
AK297401.1 | - | 29 | 3715 | |||
AK303702.1 | - | 30 | 3858 | |||
AL136549.1 | - | 31 | 3982 | |||
AL161999.1 | - | 23 | 2943 | |||
BC011762.1 | - | 31 | 4109 | 3619680 | ||
BC021008.1 | - | 31 | 4109 | |||
L47738.1 | - | 20 | 2756 |