Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000615.6 | NP_000606.3 | 4 | 724 | ||
NM_001076682.3 | NP_001070150.1 | 4 | 724 | |||
NM_001242607.1 | NP_001229536.1 | 4 | 724 | |||
NM_001242608.1 | NP_001229537.1 | 4 | 724 | |||
NM_181351.4 | NP_851996.2 | 4 | 724 | |||
GenBank mRNA | AB209443.1 | - | 4 | 603 | ||
AF005070.1 | - | 4 | 570 | |||
AK057509.1 | - | 4 | 827 | |||
AK292453.1 | - | 4 | 359 | |||
AK314589.1 | - | 4 | 696 | |||
BC029119.1 | - | 4 | 535 | |||
BC047244.1 | - | 4 | 536 | 5590188 | ||
S71824.1 | - | 4 | 559 | |||
U63041.1 | - | 4 | 395 | |||
X16841.1 | - | 4 | 503 |